2,5-Difluoro-4-(trifluoromethyl)benzoic acid 99%

Lieferant: Apollo Scientific

PC907130-250MG PC907130-1G PC907130-5G
APOSPC907130-250MGEA 0 EUR
APOSPC907130-250MG APOSPC907130-1G APOSPC907130-5G
2,5-Difluoro-4-(trifluoromethyl)benzoic acid 99%
2,5-Difluoro-4-(trifluoromethyl)benzoic acid
Formel: C₈H₃F₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01631618
CAS-Nummer: 261945-05-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR