7-Fluoro-6-methyl isatin 95%

Lieferant: Apollo Scientific

PC907136-1G
APOSPC907136-1GEA 647 EUR
APOSPC907136-1G
7-Fluoro-6-methyl isatin 95%
7-Fluoro-6-methyl isatin
Formel: C₉H₆FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11042558
CAS-Nummer: 1073262-83-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR