2,3-Difluoro-6-(trifluoromethyl)aniline 95%

Lieferant: Apollo Scientific

PC907139-1G
APOSPC907139-1GEA 310 EUR
APOSPC907139-1G
2,3-Difluoro-6-(trifluoromethyl)aniline 95%
2,3-Difluoro-6-(trifluoromethyl)aniline
Formel: C₇H₄F₅N
Molecular Weight: 197,105 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07777167
CAS-Nummer: 124185-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR