Methyl 3,6-difluoro-2-hydroxybenzoate 95%

Lieferant: Apollo Scientific

PC907140-1G
APOSPC907140-1GEA 647 EUR
APOSPC907140-1G
Methyl 3,6-difluoro-2-hydroxybenzoate 95%
Methyl 3,6-difluoro-2-hydroxybenzoate
Formel: C₈H₆F₂O₃
Molecular Weight: 188,128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10567189
CAS-Nummer: 1214324-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR