4-Chloro-5-fluoroindoline 95%

Lieferant: Apollo Scientific

PC907195-1G
APOSPC907195-1GEA 806 EUR
APOSPC907195-1G
4-Chloro-5-fluoroindoline 95%
4-Chloro-5-fluoroindoline
Formel: C₈H₇ClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD12913309
CAS-Nummer: 903551-32-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR