2-(2,3-Difluorophenyl)acetaldehyde 95%
Lieferant: Apollo Scientific
PC907200-1G
APOSPC907200-1GEA
1260
EUR
APOSPC907200-1G
2-(2,3-Difluorophenyl)acetaldehyde 95%
2-(2,3-Difluorophenyl)acetaldehyde
Formel:
C₈H₆F₂O Molecular Weight: 156.129 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11553568 CAS-Nummer: 866342-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...