7-Fluorobenzo[d]thiazol-2-amine 95%
Lieferant: Apollo Scientific
PC907203-1G
APOSPC907203-1GEA
562
EUR
APOSPC907203-1G
7-Fluorobenzo[d]thiazol-2-amine 95%
7-Fluorobenzo[d]thiazol-2-amine
Formel:
C₇H₅FN₂S Molecular Weight: 168,191 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11110958 CAS-Nummer: 20358-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...