3-Chloro-2-fluoro-5-iodobenzoic acid ≥95%

Lieferant: Apollo Scientific

PC907221-250MG
APOSPC907221-250MGEA 115 EUR
APOSPC907221-250MG
3-Chloro-2-fluoro-5-iodobenzoic acid ≥95%
3-Chloro-2-fluoro-5-iodobenzoic acid
Formel: C₇H₃ClFIO₂
Molecular Weight: 300,453 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD24107275
CAS-Nummer: 1000162-09-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR