4,5-Difluoro-2-iodoaniline 95%

Lieferant: Apollo Scientific

PC907253-5G
APOSPC907253-5GEA 188 EUR
APOSPC907253-5G
4,5-Difluoro-2-iodoaniline 95%
4,5-Difluoro-2-iodoaniline
Formel: C₆H₄F₂IN
Molecular Weight: 255.004 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095321
CAS-Nummer: 847685-01-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR