4-Fluoro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one 97%

Lieferant: Apollo Scientific
Achtung

PC907269-250MG
APOSPC907269-250MGEA 168 EUR
APOSPC907269-250MG
4-Fluoro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one 97%
4-Fluoro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one
Formel: C₇H₅FN₂O
Lagertemperatur: Kühlschrank
MDL: MFCD12923876
CAS-Nummer: 956460-93-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR