2-Chloro-4-(difluoromethyl)pyrimidine ≥98%
Lieferant: Apollo Scientific
PC907312-100MG
APOSPC907312-100MGEA
116
EUR
APOSPC907312-100MG
2-Chloro-4-(difluoromethyl)pyrimidine ≥98%
2-Chloro-4-(difluoromethyl)pyrimidine
Formel:
C₅H₃ClF₂N₂ Molecular Weight: 164,541 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18415621 CAS-Nummer: 1261629-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...