5-(Trifluoromethyl)isoindoline 95%

Lieferant: Apollo Scientific

PC907326-1G
APOSPC907326-1GEA 2430 EUR
APOSPC907326-1G
5-(Trifluoromethyl)isoindoline 95%
5-(Trifluoromethyl)isoindoline
Formel: C₉H₈F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD08234859
CAS-Nummer: 342638-03-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR