2-(Trifluoromethyl)furan-3-carboxylic acid 95+%

Lieferant: Apollo Scientific
Achtung

PC907334-250MG
APOSPC907334-250MGEA 111 EUR
APOSPC907334-250MG
2-(Trifluoromethyl)furan-3-carboxylic acid 95+%
2-(Trifluoromethyl)furan-3-carboxylic acid
Formel: C₆H₃F₃O₃
Molecular Weight: 180,081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03662621
CAS-Nummer: 536718-30-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR