(3-Fluorocyclobutyl)methanol

Lieferant: Apollo Scientific

PC907389-1G
APOSPC907389-1GEA 1150 EUR
APOSPC907389-1G
(3-Fluorocyclobutyl)methanol
(3-Fluorocyclobutyl)methanol
Formel: C₅H₉FO
Lagertemperatur: Raumtemperatur
MDL: MFCD15527038
CAS-Nummer: 1260654-20-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR