A-Trifluoroacetyl-g-butyrolactone 90%

Lieferant: Apollo Scientific

PC907493-1G
APOSPC907493-1GEA 647 EUR
APOSPC907493-1G
A-Trifluoroacetyl-g-butyrolactone 90%
A-Trifluoroacetyl-g-butyrolactone
Formel: C₆H₅F₃O₃
Molecular Weight: 182,097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00525555
CAS-Nummer: 69243-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR