2-(Difluoromethoxy)-6-fluorobenzonitrile 97%

Lieferant: Apollo Scientific

PC907519-1G
APOSPC907519-1GEA 1330 EUR
APOSPC907519-1G
2-(Difluoromethoxy)-6-fluorobenzonitrile 97%
2-(Difluoromethoxy)-6-fluorobenzonitrile
Formel: C₈H₄F₃NO
Molecular Weight: 187,119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18394953
CAS-Nummer: 221202-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR