2-(2-(Trifluoromethyl)phenyl)acetamide 95%

Lieferant: Apollo Scientific

PC907521-25G
APOSPC907521-25GEA 85.7 EUR
APOSPC907521-25G
2-(2-(Trifluoromethyl)phenyl)acetamide 95%
2-(2-(Trifluoromethyl)phenyl)acetamide
Formel: C₉H₈F₃NO
Molecular Weight: 203,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153288
CAS-Nummer: 34779-65-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR