(2-Amino-5-fluorophenyl)(phenyl)methanone 95%

Lieferant: Apollo Scientific

PC907525-1G
APOSPC907525-1GEA 1320 EUR
APOSPC907525-1G
(2-Amino-5-fluorophenyl)(phenyl)methanone 95%
(2-Amino-5-fluorophenyl)(phenyl)methanone
Formel: C₁₃H₁₀FNO
Molecular Weight: 215,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00179567
CAS-Nummer: 362-46-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR