Methyl-2-chloro-6-(trifluoromethyl)nicotinate

Lieferant: Apollo Scientific

PC907776-1G PC907776-250MG PC907776-5G
APOSPC907776-1GEA 0 EUR
APOSPC907776-1G APOSPC907776-250MG APOSPC907776-5G
Methyl-2-chloro-6-(trifluoromethyl)nicotinate
Methyl-2-chloro-6-(trifluoromethyl)nicotinate
Formel: C₈H₅ClF₃NO₂
Lagertemperatur: Kühlschrank
MDL: MFCD14698016
CAS-Nummer: 1073129-57-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR