2-Chloro-5-fluoro-N-methylpyrimidin-4-amine 95%

Lieferant: Apollo Scientific

PC907811-1G PC907811-250MG
APOSPC907811-1GEA 754 EUR
APOSPC907811-1G APOSPC907811-250MG
2-Chloro-5-fluoro-N-methylpyrimidin-4-amine 95%
2-Chloro-5-fluoro-N-methylpyrimidin-4-amine
Formel: C₅H₅ClFN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01346699
CAS-Nummer: 67316-43-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR