(1R)-1-(3-Fluorophenyl)ethylamine 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC9079-1G
PC9079-250MG
APOSPC9079-1GEA
109
EUR
APOSPC9079-1G
APOSPC9079-250MG
(1R)-1-(3-Fluorophenyl)ethylamine 97%
Formel:
C₈H₁₀FN Flammpunkt: 72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06761859 CAS-Nummer: 761390-58-3 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...