7-Fluoroisoquinoline ≥98%
Lieferant: Apollo Scientific
PC907907-100MG
PC907907-1G
APOSPC907907-100MGEA
85.7
EUR
APOSPC907907-100MG
APOSPC907907-1G
7-Fluoroisoquinoline ≥98%
7-Fluoroisoquinoline
Formel:
C₉H₆FN Molecular Weight: 147.149 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226905 CAS-Nummer: 1075-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...