7-Fluoroisoquinoline ≥98%

Lieferant: Apollo Scientific

PC907907-100MG PC907907-1G
APOSPC907907-100MGEA 85.7 EUR
APOSPC907907-100MG APOSPC907907-1G
7-Fluoroisoquinoline ≥98%
7-Fluoroisoquinoline
Formel: C₉H₆FN
Molecular Weight: 147.149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226905
CAS-Nummer: 1075-12-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR