N-(2-(Ethylamino)ethyl)-2,2,2-trifluoroacetamide 95%

Lieferant: Apollo Scientific

PC907919-1G
APOSPC907919-1GEA 647 EUR
APOSPC907919-1G
N-(2-(Ethylamino)ethyl)-2,2,2-trifluoroacetamide 95%
N-(2-(Ethylamino)ethyl)-2,2,2-trifluoroacetamide
Formel: C₆H₁₁F₃N₂O
Molecular Weight: 184,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18206209
CAS-Nummer: 911229-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR