2-(4-Fluorophenyl)-2-oxoacetic acid 95%

Lieferant: Apollo Scientific

PC907923-1G
APOSPC907923-1GEA 82.8 EUR
APOSPC907923-1G
2-(4-Fluorophenyl)-2-oxoacetic acid 95%
2-(4-Fluorophenyl)-2-oxoacetic acid
Formel: C₈H₅FO₃
Molecular Weight: 168,122 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02250293
CAS-Nummer: 2251-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR