3-Fluoro-N-methyl-4-nitrobenzamide 95%

Lieferant: Apollo Scientific

PC907963-1G
APOSPC907963-1GEA 203 EUR
APOSPC907963-1G
3-Fluoro-N-methyl-4-nitrobenzamide 95%
3-Fluoro-N-methyl-4-nitrobenzamide
Formel: C₈H₇FN₂O₃
Molecular Weight: 198,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14583038
CAS-Nummer: 658700-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR