2-Fluor-4'-chloracetophenon 95%
Lieferant: Apollo Scientific
Synonyme:
1-(4-Chlorophenyl)-2-fluoroethanone
PC907972-1G
APOSPC907972-1GEA
440
EUR
APOSPC907972-1G
2-Fluor-4'-chloracetophenon 95%
2-Fluor-4'-chloracetophenon
Formel:
C₈H₆ClFO Molecular Weight: 172,5840 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03412196 CAS-Nummer: 329-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...