2-(2-Chloro-6-fluorophenyl)ethanol 95%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-6-fluorophenethyl alcohol

PC907978-25G
APOSPC907978-25GEA 909 EUR
APOSPC907978-25G
2-(2-Chloro-6-fluorophenyl)ethanol 95%
2-(2-Chloro-6-fluorophenyl)ethanol
Formel: C₈H₈ClFO
Molecular Weight: 174,599 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00800609
CAS-Nummer: 214262-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR