2,5-Dichloro-3-fluorobenzoic acid 95%

Lieferant: Apollo Scientific

PC907983-25G
APOSPC907983-25GEA 970 EUR
APOSPC907983-25G
2,5-Dichloro-3-fluorobenzoic acid 95%
2,5-Dichloro-3-fluorobenzoic acid
Formel: C₇H₃Cl₂FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13250174
CAS-Nummer: 501008-42-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR