5-Chloro-2-fluoro-4-hydroxybenzaldehyde 95%

Lieferant: Apollo Scientific

PC908022-1G
APOSPC908022-1GEA 400 EUR
APOSPC908022-1G
5-Chloro-2-fluoro-4-hydroxybenzaldehyde 95%
5-Chloro-2-fluoro-4-hydroxybenzaldehyde
Formel: C₇H₄ClFO₂
Molecular Weight: 174,557 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22066584
CAS-Nummer: 838856-31-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR