Methyl 5-amino-2-fluoro-4-methylbenzoate 95%

Lieferant: Apollo Scientific

PC908033-1G
APOSPC908033-1GEA 159 EUR
APOSPC908033-1G
Methyl 5-amino-2-fluoro-4-methylbenzoate 95%
Methyl 5-amino-2-fluoro-4-methylbenzoate
Formel: C₉H₁₀FNO₂
Molecular Weight: 183,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23863661
CAS-Nummer: 1504965-88-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR