1-(2-Fluoro-6-methylphenyl)ethanone 95%

Lieferant: Apollo Scientific

PC908040-1G
APOSPC908040-1GEA 464 EUR
APOSPC908040-1G
1-(2-Fluoro-6-methylphenyl)ethanone 95%
1-(2-Fluoro-6-methylphenyl)ethanone
Formel: C₉H₉FO
Molecular Weight: 152.166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194344
CAS-Nummer: 1214366-42-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR