2-Cyano-5-fluorobenzyl chloride 95%

Lieferant: Apollo Scientific

Synonyme: 2-(Chloromethyl)-4-fluorobenzonitrile

PC908042-1G
APOSPC908042-1GEA 287 EUR
APOSPC908042-1G
2-Cyano-5-fluorobenzyl chloride 95%
2-Cyano-5-fluorobenzyl chloride
Formel: C₈H₅ClFN
Molecular Weight: 169,583 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18394047
CAS-Nummer: 1261742-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR