4-(2-Fluoro-1-fluoromethyl-ethoxy)-aniline 95%

Lieferant: Apollo Scientific

PC908061-1G
APOSPC908061-1GEA 647 EUR
APOSPC908061-1G
4-(2-Fluoro-1-fluoromethyl-ethoxy)-aniline 95%
4-(2-Fluoro-1-fluoromethyl-ethoxy)-aniline
Formel: C₉H₁₁F₂NO
Molecular Weight: 187,187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972874
CAS-Nummer: 109230-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR