5,6-Difluoro-1H-indazole 95+%

Lieferant: Apollo Scientific
Achtung

PC908143-250MG
APOSPC908143-250MGEA 219 EUR
APOSPC908143-250MG
5,6-Difluoro-1H-indazole 95+%
5,6-Difluoro-1H-indazole
Formel: C₇H₄F₂N₂
Molecular Weight: 154,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09997804
CAS-Nummer: 944898-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR