3-Fluorocyclobutane-1-carbonitrile

Lieferant: Apollo Scientific

PC908417-1G
APOSPC908417-1GEA 1810 EUR
APOSPC908417-1G
3-Fluorocyclobutane-1-carbonitrile
3-Fluorocyclobutane-1-carbonitrile
Formel: C₅H₆FN
Molecular Weight: 99,106 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26727206
CAS-Nummer: 1552638-51-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR