3-(Trifluoroacetyl)pyridine 95%
Lieferant: Apollo Scientific
Synonyme:
2,2,2-Trifluoro-1-(pyridin-3-yl)ethanol
PC908420-1G
PC908420-250MG
APOSPC908420-1GEA
806
EUR
APOSPC908420-1G
APOSPC908420-250MG
3-(Trifluoroacetyl)pyridine 95%
3-(Trifluoroacetyl)pyridine
Formel:
C₇H₆F₃NO Molecular Weight: 177.126 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16067922 CAS-Nummer: 138624-99-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...