5-Fluoroimidazo[1,2-a]pyridine hydrochloride 95%

Lieferant: Apollo Scientific

PC908467-1G
APOSPC908467-1GEA 734 EUR
APOSPC908467-1G
5-Fluoroimidazo[1,2-a]pyridine hydrochloride 95%
5-Fluoroimidazo[1,2-a]pyridine hydrochloride
Formel: C₇H₅FN₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD08705707
CAS-Nummer: 198896-14-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR