6-Fluoro-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid 95%

Lieferant: Apollo Scientific

PC908509-1G
APOSPC908509-1GEA 299 EUR
APOSPC908509-1G
6-Fluoro-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid 95%
6-Fluoro-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid
Formel: C₁₀H₁₀FNO₂
Molecular Weight: 195,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114844
CAS-Nummer: 1260641-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR