2-Chloro-7-fluoro-1,5-naphthyridine ≥97%
Lieferant: Apollo Scientific
PC908524-250MG
APOSPC908524-250MGEA
159
EUR
APOSPC908524-250MG
2-Chloro-7-fluoro-1,5-naphthyridine ≥97%
2-Chloro-7-fluoro-1,5-naphthyridine
Formel:
C₈H₄ClFN₂ Molecular Weight: 182.582 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD19687920 CAS-Nummer: 1378470-92-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...