5-Fluor-8-methoxychinolin 95%

Lieferant: Apollo Scientific

PC908543-1G
APOSPC908543-1GEA 71.2 EUR
APOSPC908543-1G
5-Fluor-8-methoxychinolin 95%
5-Fluor-8-methoxychinolin
Formel: C₁₀H₈FNO
Molecular Weight: 177,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18452297
CAS-Nummer: 439-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR