5-Amino-2-(trifluoromethyl)benzonitrile

Lieferant: Apollo Scientific
Achtung

PC908568-1G
APOSPC908568-1GEA 0 EUR
APOSPC908568-1G
5-Amino-2-(trifluoromethyl)benzonitrile
5-Amino-2-(trifluoromethyl)benzonitrile
Formel: C₈H₅F₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD20694851
CAS-Nummer: 354814-19-0
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR