5-Chloro-6-(trifluoromethyl)pyridin-2-amine 95%
Lieferant: Apollo Scientific
PC908580-1G
APOSPC908580-1GEA
128
EUR
APOSPC908580-1G
5-Chloro-6-(trifluoromethyl)pyridin-2-amine 95%
5-Chloro-6-(trifluoromethyl)pyridin-2-amine
Formel:
C₆H₄ClF₃N₂ Molecular Weight: 196,558 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16606147 CAS-Nummer: 1227595-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...