5-Fluoro-1-methyl-1H-indole-2,3-dione 95%

Lieferant: Apollo Scientific

Synonyme: 5-Fluoro-1-methylindoline-2,3-dione

PC908631-1G
APOSPC908631-1GEA 116 EUR
APOSPC908631-1G
5-Fluoro-1-methyl-1H-indole-2,3-dione 95%
5-Fluoro-1-methyl-1H-indole-2,3-dione
Formel: C₉H₆FNO₂
Molecular Weight: 179,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02956480
CAS-Nummer: 773-91-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR