4-Chlor-2-fluor-5-methoxybenzaldehyd 95%
Lieferant: Apollo Scientific
PC908646-1G
APOSPC908646-1GEA
538
EUR
APOSPC908646-1G
4-Chlor-2-fluor-5-methoxybenzaldehyd 95%
4-Chloro-2-fluoro-5-methoxybenzaldehyde
Formel:
C₈H₆ClFO₂ Molecular Weight: 188,5800 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05863886 CAS-Nummer: 177034-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...