3-(3-Fluorophenoxy)propylamine 95%

Lieferant: Apollo Scientific

PC908667-1G PC908667-5G
APOSPC908667-1GEA 0 EUR
APOSPC908667-1G APOSPC908667-5G
3-(3-Fluorophenoxy)propylamine 95%
3-(3-Fluorophenoxy)propylamine
Formel: C₉H₁₂FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09741966
CAS-Nummer: 116753-49-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR