2-Cyan-4'-fluoracetanilid 95%

Lieferant: Apollo Scientific

Synonyme: 2-Cyano-N-(4-fluorophenyl)acetamide

PC908683-5G
APOSPC908683-5GEA 75.4 EUR
APOSPC908683-5G
2-Cyan-4'-fluoracetanilid 95%
2-Cyan-4'-fluoracetanilid
Formel: C₉H₇FN₂O
Molecular Weight: 178,1631 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00276110
CAS-Nummer: 1735-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR