N-Methyl-3-(trifluoromethyl)benzylamine 95%

Lieferant: Apollo Scientific

PC908684-5G
APOSPC908684-5GEA 125 EUR
APOSPC908684-5G
N-Methyl-3-(trifluoromethyl)benzylamine 95%
N-Methyl-3-(trifluoromethyl)benzylamine
Formel: C₉H₁₀F₃N
Molecular Weight: 189,181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115410
CAS-Nummer: 90390-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR