2-(4-Methylpiperazin-1-yl)-5-(trifluoromethyl)aniline 98%

Lieferant: Apollo Scientific
Achtung

PC908703-1G PC908703-250MG PC908703-5G
APOSPC908703-1GEA 229 EUR
APOSPC908703-1G APOSPC908703-250MG APOSPC908703-5G
2-(4-Methylpiperazin-1-yl)-5-(trifluoromethyl)aniline 98%
2-(4-Methylpiperazin-1-yl)-5-(trifluoromethyl)aniline
Formel: C₁₂H₁₆F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03848692
CAS-Nummer: 685853-98-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR