3-Amino-6-fluor-1H-indazol 97%

Lieferant: Apollo Scientific
Achtung

PC908712-1G PC908712-250MG PC908712-5G
APOSPC908712-1GEA 60.9 EUR
APOSPC908712-1G APOSPC908712-250MG APOSPC908712-5G
3-Amino-6-fluor-1H-indazol 97%
3-Amino-6-fluoro-1H-indazole
Formel: C₇H₆FN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06739129
CAS-Nummer: 404827-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR